Anti-CDH23 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15498R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CDH23
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:51-150/3354
- Form:Liquid
- Antigen synonyms:Mdfw|USH 1D|CDH 23|nmf252|USH1 D|Bob|Ahl1|Cadherin 23|Cadherin23|Bobby|Bus|nmf112|USH1D|Modifier of deaf waddler|Waltzer|Ahl|Bustling|CAD23_HUMAN|nmf181|Ahl 1|Age related hearing loss 1|Otocadherin
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:367 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CDH23
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
CDH23.
Type: Primary
Antigen: CDH23
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/3354
Host: Rabbit
Isotype: IgG
Reactivity: Human