Anti-HABP4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15402R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HABP4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:41-140/413
- Form:Liquid
- Antigen synonyms:HABP4_HUMAN|IHABP-4|IHABP4|Intracellular hyaluronan-binding protein 4|Ki-1/57 intracellular antigen
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:46 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HABP4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
HABP4.
Type: Primary
Antigen: HABP4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-140/413
Host: Rabbit
Isotype: IgG
Reactivity: Mouse