Anti-GPCR TGR7 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Catalog # BOSSBS-15374R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GPCR TGR7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-200/321
- Form:Liquid
- Antigen synonyms:Beta-alanine receptor|Mas-related G-protein coupled receptor member D|MRGPRD|G-protein coupled receptor TGR7|MRGD|TGR7|MRGRD|MRGRD_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:36 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPCR TGR7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GPCR TGR7.
Type: Primary
Antigen: GPCR TGR7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 101-200/321
Host: Rabbit
Isotype: IgG
Reactivity: