Anti-C7ORF72 Rabbit Polyclonal Antibody (Cy7)
BOSSBS-15282R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:C7orf72
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:261-360/438
- Form:Liquid
- Antigen synonyms:CG072_HUMAN|Chromosome 7 open reading frame 72|Uncharacterized protein C7orf72|C7orf72
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:50 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C7ORF72
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C7orf72.
Type: Primary
Antigen: C7orf72
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 261-360/438
Host: Rabbit
Isotype: IgG
Reactivity: