Order Entry
Sweden
ContactUsLinkComponent
Anti-MSY2/YBOX2/DBPC Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Anti-MSY2/YBOX2/DBPC Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Catalog # BOSSBS-12271R-A750
Supplier:  Bioss
Anti-MSY2/YBOX2/DBPC Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Catalog # BOSSBS-12271R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MSY2/YBOX2/DBPC
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    101-185/364
  • Form:
    Liquid
  • Gene ID:
    51087
  • Antigen synonyms:
    DBPC|Germ cell-specic Y-box-binding protein|Y-box-binding protein 2|OTTMUSP00000006276|MSY2 homolog|Contrin|YBOX2_HUMAN|FRGY2 homolog|MGC118270|Germ cell specic Y box binding protein|MGC45104|YBX 2|YBX2|MSY2|CSDA 3|Y box binding protein 2|MSY 2|CSDA3|DNA binding protein C|RGD1305068|DNA-binding protein C
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human YBOX2
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

MSY2 and YB-2 (MSY3,4) belong to the Y-box family of multifunctional proteins that regulate both transcription and translation (13). Y-box proteins interact with a wide variety of nucleic acid structures to act as transcription factors and mRNA masking proteins (1). The modular structure of Y-box proteins includes a highly conserved N-terminal cold-shock domain (CSD, equivalent to the bacterial cold-shock proteins) and four basic C-terminal domains containing arginine clusters and aromatic residues (4). MSY2 is expressed in testis and ovary where it may repress translation of parental mRNA (5,6). The gene encoding human MSY2 maps to chromosome 17p11.2-13.1 (5). YB-2 (MSY3,4 in mouse) is also known as DNA binding protein A and is highly expressed in the testis, heart and muscle (7,8). MSY2 and YB-2 bind to the consensus sequence 5'-UCCAUCA-3' contained in the Y-box element (9).

Type: Primary
Antigen: MSY2/YBOX2/DBPC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-185/364
Host: Rabbit
Isotype: IgG
Reactivity: