About this item
RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.
- For maintenance and propagation of E. Coli
- High salt formula
- Molecular biology grade
Simply dissolve 1 capsule per 1 L of distilled water. Water must be 37 °C or warmer to completely dissolve gelatin capsule. Agitate. Sterilise by autoclaving for 15 minutes before use. Buffer is set for 7,2 at 20 °C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.
Microorganism (E. coli ATCC 23724): Good to Excellent
Microorganism (E. coli ATCC 33694): Good to Excellent
Microorganism (E. coli ATCC 33849): Fair to Excellent
Microorganism (E. coli ATCC 39403): Good to Excellent
Microorganism (E. coli ATCC 47014): Good to Excellent
Microorganism (E. coli ATCC 53868): Good to Excellent
Specifications
- Format:Capsules
- Form:Cream to pale beige free flowing powder
- pH:6,8 - 7,2 (2,5 % Solution at 25 °C)