Anti-Ctip1 Rabbit Polyclonal Antibody
Catalog # NOVUNB600-261
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:Ctip1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:53335
- Antigen synonyms:EVI-9|B-cell CLL/lymphoma 11A|B-cell CLL/lymphoma 11A (zinc finger protein)|Zinc finger protein 856|B-cell lymphoma/leukemia 11A|C2H2-type zinc finger protein|BCL11A-XL|KIAA1809BCL11A-L|BCL-11A|COUP-TF-interacting protein 1|ZNF856FLJ10173|HBFQTL5|B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2|CTIP1ecotropic viral integration site 9|EVI9FLJ34997|BCL11A-S|BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1|ecotropic viral integration site 9 homolog|Ecotropic viral integration site 9 protein homolog
- Storage buffer:Tris-Citrate/Phosphate (pH 7,0 - 8,0)
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:1,0 mg/ml
- Immunogen:A synthetic peptide which maps to a region between residues 775 and the C-terminus (residue 835) of human B-cell CLL/Lymphoma 11A using the numbering given in Swiss-Prot entry Q9H165 (GeneID 53335).
- Purification:Immunogen affinity purified
- Pk:100 µl
Specifications
About this item
The Ctip1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ctip1. This antibody reacts with human, mouse. The Ctip1 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunoprecipitation.
There is 1 scientific publication usiµg this antibody.
Type: Primary
Antigen: Ctip1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse