Order Entry
Sweden
ContactUsLinkComponent
Glutathione Fluorometric Assay Kit
Glutathione Fluorometric Assay Kit
Catalog # BIOVK251-100
Supplier:  Abcam
CAS Number:  
undefined
Glutathione Fluorometric Assay Kit
Catalog # BIOVK251-100
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    100 Assays
  • Storage conditions:
    –70 °C
  • Species reactivity:
    Mammalian samples
  • Sample type:
    Cell and tissue lysates, culture media, urine, plasma and serum, as well as many other biological fluids
  • Shelf life:
    12 Months

Specifications

About this item

Simple assay to detect total glutathione.

  • Fluorescence (Ex/Em 380/461 nm)
  • Mammalian
  • The Glutathione Detection Kit provides a simple in vitro assay for detecting the GSH changes in apoptosis and other pathological processes

Glutathione is the principal intracellular low-molecular-weight thiol that plays a critical role in the cellular defence against oxidative and nitrosative stress in mammalian cells. Diminished glutathione levels have been observed in the early stages of apoptosis. BioVision’s ApoGSH™ Glutathione Detection Kit provides a simple in vitro assay for detection of total glutathione changes in apoptosis and other samples. The assay utilizes monochlorobimane (MCB), a dye that appears to form an adduct exclusively with glutathione. The unbound MCB is almost nonfluorescent, whereas the dye fluoresces blue (Ex./Em. = 380/461 nm) when bound to glutathione of reduced or oxidized form. The reaction is catalysed by glutathione S-transferase. Thus, the amount of total glutathione can be easily detected using a fluorometer or a 96-well fluorometric plate reader.