BMS 345541 (trifluoroacetate salt) ≥98%
Supplier: Cayman Chemical
Certificates
About this item
:
Formula:C₁₄H₁₇N₅·2CF₃COOH
:483.4 g/mol
Storage Temperature:Freezer
Specification Test Results
- Formulation:A crystalline solid
- Purity:≥98%
- Stability:≥ 2 years
Specification Test Results
Frequently Bought Together