Whizard® Silver Talon Cut-Resistant Gloves
Supplier: WELLS LAMONT
Certificates
About this item
Offers a very high level of cut resistance in cleanroom environments.
- Reusable cleanroom glove
- ANSI A7 cut resistance
- Made in USA
Category III for particulates.
Certifications: ANSI A7 level F cut resistance, ANSI Level 3 abrasion resistance.
Specifications
- Catalog No:
- 112-6368
- 112-6371
- 112-6369
- 112-6370
- 112-6366
- 112-6367
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Material:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile