Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Sucrose
D-(+)-Sucrose, crystals ACS
D-(+)-Sucrose, crystals ACS
D-(+)-Sucrose, crystals ACS
Supplier:
Spectrum Chemical
Certificates
Expand image
D-(+)-Sucrose, crystals ACS
Supplier:
Spectrum Chemical
About this item
CAS Number:
57-50-1
:
Formula:
C₁₂H₂₂O₁₁
MDL Number:
MFCD00006626
:
342.3 g/mol