Order Entry
Sweden
ContactUsLinkComponent
Industrial gloves, EDGE® 48-305
Industrial gloves, EDGE® 48-305
Supplier:  Ansell
Certificates
undefined
Industrial gloves, EDGE® 48-305
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE48-305/7
  • ANSE48-305/8
  • ANSE48-305/10
  • ANSE48-305/9
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Liner material:
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Finish:
  • Crinkle grip finish (natural rubber latex)
  • Crinkle grip finish (natural rubber latex)
  • Crinkle grip finish (natural rubber latex)
  • Crinkle grip finish (natural rubber latex)
  • Gauge:
  • 10
  • 10
  • 10
  • 10
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Palm thickness:
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • EN norm:
  • EN 388:2016 (2142B)
  • EN 388:2016 (2142B)
  • EN 388:2016 (2142B)
  • EN 388:2016 (2142B)
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
Specifications