Analysis and filtration paper, Tesorb
Supplier: TERVAKOSKI OY
Certificates
About this item
An extremely pure paper perfectly manufactured for filtration, analysis and laboratory applications.
- 80 – 300 g/m²
- Made of 100% cotton linters and de-ionized process water only
- Low ash content
- No sizing, fillers or bindings
- High content of α-cellulose
- Consistent performance
100% de-ionized process water guarantees extremely pure quality.
Frequently Bought Together