Saccharin ≥99.0%, powder NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:81-07-2
:
Formula:C₇H₅NO₃S
MDL Number:MFCD00005866
:183.19 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (C₇H₅NO₃S):99.0 - 101.0 %
- Clarity of Solution:To Pass Test
- Color of Solution:To Pass Test
- Melting Range:226 - 230 °C
- Loss On Drying:max. 1.0 %