Petrolatum, white USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:8009-03-8
:
MDL Number:MFCD00147839
Storage Temperature:Ambient
Specification Test Results
- BHT:As Reported
- Tocopherols:As Reported
- Specific Gravity @60 C:0.815 - 0.880
- Melting Range:38 - 60 °C