Thermoplastic duct
: LABCONCO
PVC exhaust duct is type one, unplasticised, schedule 40, lightweight, and corrosion-resistant.
A female duct coupling is required to join two sections. Connections can be made using solvent cement. PVC is rigid and may be cut without the use of special tools.