Order Entry
Sweden
ContactUsLinkComponent
2,2,4-Trimethylpentane, liquid, Burdick & Jackson™
2,2,4-Trimethylpentane, liquid, Burdick & Jackson™
Catalog # HONC65722-2.5L
CAS Number:  
2,2,4-Trimethylpentane, liquid, Burdick & Jackson™
Catalog # HONC65722-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :114.23 g/mol
Boiling Pt:99 °C (1,013 hPa)
Melting Pt:-107 °C
Melting Pt:–107 °C
Density:0.69 g/cm3 (20 °C)
Flash Pt:-12 °C
UN:1262
ADR:3,II
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
Flash Pt:–12 °C