Sulphosulphuron Analytical standard, reference material, Fluka™
Catalog # HONCR1986-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:141776-32-1
- Description:Sulphosulphuron
- MDL No.:MFCD04034670
- Synonym(s):Sulfosulfuron; 1-(4,6-Dimethoxypyrimidin-2-yl)-3-(2-ethylsulfonylimidazol[1,2-a]pyridin-3-ylsulfonyl)urea; N-[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]-2-(ethylsulfonyl)imidazo[1,2-a]pyridine-3-sulfonamide
- Melting point:198 °C
- Formula:C₁₆H₁₈N₆O₇S₂
- Molecular weight:470.48 g/mol
- Description:Sulphosulphuron
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery