Isoproturon Analytical standard, reference material, Fluka™
Catalog # HONCR1977-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:34123-59-6
- Description:Isoproturon
- MDL No.:MFCD00078684
- Synonym(s):3-(4-Isopropylphenyl)-1,1-dimethylurea
- Melting point:155 °C
- Formula:C₁₂H₁₈N₂O
- Molecular weight:206.28 g/mol
- Description:Isoproturon
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery