Imazapyr Analytical standard, reference material, Fluka™
HONCR1970-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:81334-34-1
- Description:Imazapyr
- MDL No.:MFCD00144470
- Synonym(s):2-[(4RS)-4-isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl]pyridine-3-carboxylic acid
- Melting point:172 - 173 °C
- Formula:C₁₃H₁₅N₃O₃
- Molecular weight:261.28 g/mol
- Description:Imazapyr
- Physical form:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery