Isopentyl acetate Analytical standard, reference material, Fluka™
Catalog # HONCR1934-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:123-92-2
- Description:Isopentyl acetate
- Storage conditions:RT
- MDL No.:MFCD00008946
- UN No.:1104
- Synonym(s):Acetic acid 3-methylbutyl ester
- Formula:C₇H₁₄O₂
- Molecular weight:130.18 g/mol
- Description:Isopentyl acetate
- Flash point:25 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery