1-Pentanol Analytical standard, reference material, Fluka™
Katalog # HONCR1930-1G
Leverantör: Honeywell Chemicals
CAS-nummer:
Specifikationer
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:71-41-0
- Description:1-Pentanol
- Storage conditions:RT
- MDL No.:MFCD00002977
- UN No.:1105
- Synonym(s):n-Amyl alcohol
- Formula:C₅H₁₂O
- Molecular weight:88.15 g/mol
- Description:1-Pentanol
- Flash point:49 °C
- Physical form:Liquid
Specifikationer
Om denna produkt
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery