2-Butanol Analytical standard, reference material, Fluka™
Catalog # HONCR1924-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:78-92-2
- Description:2-Butanol
- Storage conditions:RT
- MDL No.:MFCD00004569
- UN No.:1120
- Synonym(s):sec-Butyl alcohol
- Melting point:-89 °C
- Formula:C₄H₁₀O
- Molecular weight:74.12 g/mol
- Description:2-Butanol
- Flash point:24 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery