Ethyl benzoate Analytical standard, reference material, Fluka™
Catalog # HONCR1916-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:93-89-0
- Description:Ethyl benzoate
- Storage conditions:RT
- MDL No.:MFCD00009109
- Synonym(s):Ethyl benzenecarboxylate
- Melting point:-33 °C
- Formula:C₉H₁₀O₂
- Molecular weight:150.18 g/mol
- Description:Ethyl benzoate
- Flash point:closed cup.88 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery