Ethyl heptanoate Analytical standard, reference material, Fluka™
Catalog # HONCR1908-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:106-30-9
- Description:Ethyl heptanoate
- Storage conditions:RT
- MDL No.:MFCD00009538
- Synonym(s):Heptanoic acid ethyl ester; Ethyl enanthate
- Melting point:-65 °C
- Formula:C₉H₁₈O₂
- Molecular weight:158.24 g/mol
- Description:Ethyl heptanoate
- Flash point:70.5 °C.1,013 hPa
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery