Allyl alcohol Analytical standard, reference material, Fluka™
Catalog # HONCR1905-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:107-18-6
- Description:Allyl alcohol
- Storage conditions:RT
- MDL No.:MFCD00002920
- UN No.:1098
- Synonym(s):2-Propen-1-ol
- Melting point:-129 °C
- Formula:C₃H₆O
- Molecular weight:58.07 g/mol
- Description:Allyl alcohol
- Flash point:21 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery