Chloramphenicol-solution, 1mg/L in acetonitrile Analytical standard, reference material, Fluka™
HONCR1901-1ML
:
- Pack type:Glass Bottle
- Pk:1 mL
- Standards:Analytical standard
- Description:Chloramphenicol-solution, 1mg/L in acetonitrile
- Storage conditions:Ambient
- MDL No.:MFCD00078159
- UN No.:1648
- Synonym(s):D-(-)-threo-2,2-Dichloro-N-[ß-hydroxy-a-(hydroxymethyl)-ß-(4-nitrophenyl)ethyl]acetamide
- Melting point:-46 °C.Data refer to the solvent.
- Concentration:1mg/L in acetonitrile
- Description:Chloramphenicol
- Flash point:6 °C.The information regarding the flash point is that of the solvent.
- Physical form:Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery