Flupyradifurone Analytical standard, reference material, Fluka™
Catalog # HONCR1898-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:951659-40-8
- Description:Flupyradifurone
- Storage conditions:RT
- MDL No.:MFCD22373627
- Synonym(s):4-[[(6-Chloro-3-pyridinyl)methyl](2,2-difluoroethyl)amino]-2(5H)-furanone
- Melting point:69 °C
- Formula:C₁₂H₁₁ClF₂N₂O₂
- Molecular weight:288.7 g/mol
- Description:Flupyradifurone
- Flash point:100 °C
- Physical form:Powder
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery