Metazachlor Analytical standard, reference material, Fluka™
Catalog # HONCR1895-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:67129-08-2
- Description:Metazachlor
- Storage conditions:RT
- MDL No.:MFCD00055509
- Synonym(s):2-Chloro-N-(2,6-dimethylphenyl)-N-(1H-pyrazol-1-ylmethyl)acetamide
- Melting point:81 °C
- Formula:C₁₄H₁₆ClN₃O
- Molecular weight:277.75 g/mol
- Description:Metazachlor
- Physical form:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery