Coumaphos Analytical standard, reference material, Fluka™
Catalog # HONCR1890-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:56-72-4
- Description:Coumaphos
- Storage conditions:2-8°C
- MDL No.:MFCD00041820
- UN No.:2811
- Synonym(s):O-(3-Chloro-4-methyl-2-oxo-2H-1-benzopyran-7-yl) O,O-diethyl phosphorothioate
- Melting point:95 °C
- Formula:C₁₄H₁₆ClO₅PS
- Molecular weight:362.77 g/mol
- Description:Coumaphos
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery