Order Entry
Sweden
ContactUsLinkComponent
 
Cefalexin Analytical standard, reference material, Fluka™
  HONCR1874-100MG
 :  
Cefalexin Analytical standard, reference material, Fluka™
  HONCR1874-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    23325-78-2
  • Description:
    Cefalexin
  • Storage conditions:
    RT
  • MDL No.:
    MFCD00056877
  • Synonym(s):
    7-(D-α-Amino-phenylacetamido)-3-methyl-3-cepheme-4-carboxylic acid
  • Formula:
    C₁₆H₁₇N₃O₄S
  • Description:
    Cefalexin
  • Physical form:
    Solid

 

 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery