Order Entry
Sweden
ContactUsLinkComponent
Triadimefon Analytical standard, reference material, Fluka™
Triadimefon Analytical standard, reference material, Fluka™
Catalog # HONCR1855-100MG
CAS Number:  
Triadimefon Analytical standard, reference material, Fluka™
Catalog # HONCR1855-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    43121-43-3
  • Description:
    Triadimefon
  • Storage conditions:
    RT
  • MDL No.:
    MFCD00055506
  • UN No.:
    2811
  • Synonym(s):
    (RS)-1-(4-chlorophenoxy)-3,3-dimethyl-1-(1H-1,2,4-triazol-1-yl)butan-2-one
  • Melting point:
    82.3 °C
  • Formula:
    C₁₄H₁₆ClN₃O₂
  • Molecular weight:
    293.75 g/mol
  • Description:
    Triadimefon
  • Flash point:
    72 °C
  • Physical form:
    Solid

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery