Order Entry
Sweden
ContactUsLinkComponent
Fenpyroximate Analytical standard, reference material, Fluka™
Fenpyroximate Analytical standard, reference material, Fluka™
Catalog # HONCR1843-100MG
CAS Number:  
Fenpyroximate Analytical standard, reference material, Fluka™
Catalog # HONCR1843-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    134098-61-6
  • Description:
    Fenpyroximate
  • Storage conditions:
    RT
  • MDL No.:
    MFCD00274596
  • UN No.:
    2811
  • Synonym(s):
    (tert-Butyl (E)-a-(1,3)-dimethyl-5-phenoxy-1H-pyrazol-4-yl methylene amino-oxy)-p-toluate)
  • Melting point:
    101.1 - 102.4 °C
  • Formula:
    C₂₄H₂₇N₃O₄
  • Molecular weight:
    421.5 g/mol
  • Description:
    Fenpyroximate
  • Physical form:
    Powder

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery