Chlorantraniliprole Analytical standard, reference material, Fluka™
HONCR1794-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:500008-45-7
- Description:Chlorantraniliprole
- Storage conditions:RT
- MDL No.:MFCD11840831
- Melting point:209 °C
- Molecular weight:483.15 g/mol
- Description:Chlorantraniliprole
- Physical form:Powder
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery