Order Entry
Sweden
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.7%, SPECTRONORM® for spectroscopy
Flammable
Irritant
tert-Butyl methyl ether ≥99.7%, SPECTRONORM® for spectroscopy
Catalog # 88441.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether ≥99.7%, SPECTRONORM® for spectroscopy
Catalog # 88441.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (GC):
    min. 99.7%
  • Water :
    max. 0.02%
  • Residue:
    max. 0.0001%
  • Colour (APHA):
    max. 10
  • Alkalinity (meq/g):
    max. 0.0002
  • Acidity (meq/g):
    max. 0.0002
  • Fluorescence (as quinine at 254 nm):
    max. 1 ppb
  • Fluorescence (as quinine at 365 nm):
    max. 1 ppb
  • UV (Transmission) (220 nm):
    min. 10%
  • UV (Transmission) (235 nm):
    min. 50%
  • UV (Transmission) (255 nm):
    min. 80%
  • UV (Transmission) (270 nm):
    min. 90%
  • UV (Transmission) (300-400 nm):
    min. 99%
  • Manufactured and filled under nitrogen:
    0

Specification Test Results

About this item

 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:−28 °C
EINECS:216-653-1
UN:2398
ADR:3,II
 :Packaged under nitrogen.
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812