Order Entry
Sweden
ContactUsLinkComponent
GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Catalog # 100-912-100
undefined
GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Catalog # 100-912-100
Specifications
  • Description:
    U.S. human serum AB
  • Seq:
    01
  • Origin:
    US
  • Storage temperature:
    Store frozen at −20…−10 °C
  • Pack type:
    Bottle
  • Pk:
    100 mL
Specifications
About this item
GemCell Plus™ Xeno-Free Human Serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.
  • Developed without the use of animal material
  • Gamma irradiation available