Pemetrexed Glutamide, Pharmaceutical Analytical Impurities
Leverantör: USP
Certifikat
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
Specifikationer
- CAS-nr:144051-68-3 (free acid)
- Synonym(s):(S)-4-((S)-4-(4-(2-(2-amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)-4-carboxybutanamido)-4-carboxybutanoate, sodium salt
- Benämning:Pemetrexed Glutamide Sodium