Atorvastatin tert-Butyl Ester, Pharmaceutical Analytical Impurities
Catalog # 1A00830.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:134395-00-9
- Description:Atorvastatin tert-Butyl Ester
- Synonym(s):tert-butyl (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate
- Description:Atorvastatin tert-Butyl Ester
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.