Order Entry
Sweden
ContactUsLinkComponent
Clopidogrel Open Ring, Pharmaceutical Analytical Impurities
Clopidogrel Open Ring, Pharmaceutical Analytical Impurities
Supplier:  USP
Certificates
Clopidogrel Open Ring, Pharmaceutical Analytical Impurities
Supplier:  USP

Specifications

  • CAS No.:
    141109-18-4
  • Synonym(s):
    methyl 2-(2-chlorophenyl)-2-((2-(thiophen-2-yl)ethyl)amino)acetate, hydrochloride
  • Description:
    Clopidogrel Open Ring Methyl Ester Hydrochloride

Specifications