Order Entry
Sweden
ContactUsLinkComponent
2-Oxo Clopidogrel, Pharmaceutical Analytical Impurities
2-Oxo Clopidogrel, Pharmaceutical Analytical Impurities
Supplier:  USP
Certificates
2-Oxo Clopidogrel, Pharmaceutical Analytical Impurities
Supplier:  USP

Specifications

  • CAS No.:
    109904-27-0
  • Synonym(s):
    methyl 2-(2-chlorophenyl)-2-(2-oxo-2,6,7,7a-tetrahydrothieno[3,2-c]pyridin-5(4H)-yl)acetate
  • Description:
    2-Oxo Clopidogrel (Mixture of Diastereomers)

Specifications