Betamethasone Valerate, Reference Standards
Leverantör: USP
Certifikat
Om denna produkt
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Betamethasone Valerate System Suitability Mixture: A mixture containing βmethasone valerate and βmethasone valerate
Certifications: US Pharmacopea Reference Standard
Specifikationer
- CAS No.:2240-28-0
- Synonym(s):9-Fluoro-11β,17-dihydroxy-16β-methyl-3,20-dioxopregna-1,4-diene-21-yl valerate
- Formula:C₂₇H₃₇FO₆
- Description:Betamethasone Valerate Related Compound A