Rivaroxaban Desmorpholinone Analog, Pharmaceutical Analytical Impurities
1A01470.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:936232-22-3
- Description:Rivaroxaban Desmorpholinone Analog
- Synonym(s):(S)-5-chloro-N-((3-(4-((2-hydroxyethyl)amino)phenyl)-2-oxooxazolidin-5-yl)methyl)thiophene-2-carboxamide
- Description:Rivaroxaban Desmorpholinone Analog
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.