Rivaroxaban Amine, Pharmaceutical Analytical Impurities
1A01160.
: USP
:
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:898543-06-1
- Description:Rivaroxaban Amine
- Synonym(s):(S)-4-(4-(5-(aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one, hydrochloride
- Description:Rivaroxaban Amine
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.