Order Entry
Sweden
ContactUsLinkComponent
 
Olanzapine, Pharmaceutical Analytical Impurities
 :  USP
 
Olanzapine, Pharmaceutical Analytical Impurities
 :  USP

 

  • CAS No.:
    1017241-34-7
  • Synonym(s):
    (Z)-4-(4-methylpiperazin-1-yl)-3-(2-oxopropylidene)-1,3-dihydro-2H-benzo[b][1,4]diazepin-2-one
  • Description:
    Olanzapine lactam