Order Entry
Sweden
ContactUsLinkComponent
Desiodoamiodarone, Pharmaceutical Analytical Impurities
Desiodoamiodarone, Pharmaceutical Analytical Impurities
Catalog # 1A00720.
Supplier:  USP
CAS Number:  
Desiodoamiodarone, Pharmaceutical Analytical Impurities
Catalog # 1A00720.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    23551-25-9
  • Description:
    Desiodoamiodarone
  • Synonym(s):
    (2-butylbenzofuran-3-yl)(4-(2-(diethylamino)ethoxy)phenyl)methanone
  • Description:
    Desiodoamiodarone

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.