Desethylamiodarone, Pharmaceutical Analytical Impurities
Catalog # 1A01080.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:96027-74-6
- Description:Desethylamiodarone
- Synonym(s):2-Butyl-3-benzofuranyl-4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl Ketone Hydrochloride
- Description:Desethylamiodarone
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.