3-Hydroxy Desloratadine, Pharmaceutical Analytical Impurities
Catalog # 1A01650.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:119410-08-1
- Description:3-Hydroxy Desloratadine
- Synonym(s):8-chloro-11-(piperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-3-ol
- Description:3-Hydroxy Desloratadine
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.