Order Entry
Sweden
ContactUsLinkComponent
Fluticasone, Pharmaceutical Analytical Impurities
Fluticasone, Pharmaceutical Analytical Impurities
Supplier:  USP
Certificates
Fluticasone, Pharmaceutical Analytical Impurities
Supplier:  USP

Specifications

  • CAS No.:
    948566-12-9
  • Synonym(s):
    (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-(propionyloxy)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carbo(thioperoxoic )O-acid
  • Description:
    Fluticasone sulfenic acid

Specifications