Order Entry
Sweden
ContactUsLinkComponent
Potassium-tert-butoxide ≥98%, GPR RECTAPUR®
Flammable
Corrosive
Potassium-tert-butoxide ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Corrosive
Potassium-tert-butoxide ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

Related Information