0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
di-Potassium hydrogen phosphate
di-Potassium hydrogen phosphate, anhydrous ≥98% for synthesis
di-Potassium hydrogen phosphate, anhydrous ≥98% for synthesis
:
VWR Chemicals
di-Potassium hydrogen phosphate, anhydrous ≥98% for synthesis
:
VWR Chemicals
:
7758-11-4
:
:
K₂HPO₄
:
MFCD00011383
:
174.18 g/mol
:
13,07743
:
340 °C
:
2.3 g/cm³ (20 °C)
:
Ambient
:
231-834-5
Assay:
Min. 98 %
SDS_EN_UK_VWRC91240.pdf
spec_VWRC91240.260
spec_VWRC91240.320